missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Click to view available options
Type:
Chlorine Reagent
Liquid Reagent Kit
Unit Size:
Each
Description
- Reagents for the colorimetric determination of free chlorine, for use with Hanna’s line of compatible benchtop and portable meters
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
- Reagents follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample
Specifications
Specifications
| Analytes | Chlorine (Free) Reagent |
| No. of Tests | 300 |
| Packaging | Dropper Bottle |
| Range | 0 to 5 mg/L |
| Test Method | Adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G |
| Type | Liquid Reagent Kit |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 C
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction