Learn More
UVP Fraud Detection Lamp
Specifications
Specifications
| Wattage | 4 W |
| Wavelength | 365 nm |
| Height (Metric) | 114 mm |
| Model | SL-2M |
| Width (Metric) | 165 mm |
| Product Type | Fraud Detection Lamp |