Learn More
FR 900098 Monosodium Salt, TRC
Product Code. 30402988
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402988 5 mg -
30395824 25 mg -
30394416 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30402988 Supplier Toronto Research Chemicals Supplier No. TRCF7635005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73226-73-0
Molecular Formula C5 H11 N O5 P . Na
Molecular Weight (g/mol) 219.11
Synonym Phosphonic acid, P-[3-(acetylhydroxyamino)propyl]-, sodium salt (1:1), Phosphonic acid, [3-(acetylhydroxyamino)propyl]-, monosodium salt (9CI), FR 900098 monosodium salt, P-[3-(Acetylhydroxyamino)propyl]phosphonic acid sodium salt, (3-(N-Hydroxyacetamido)propyl)phosphonic acid sodium salt, [3-(Acetylhydroxyamino)propyl]phosphonic acid sodium salt
IUPAC Name sodium;3-[acetyl(hydroxy)amino]propyl-hydroxyphosphinate
SMILES [Na+].CC(=O)N(O)CCCP(=O)(O)[O-]
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C5H12NO5P.Na/c1-5(7)6(8)3-2-4-12(9,10)11;/h8H,2-4H2,1H3,(H2,9,10,11);/q;+1/p-1
Formula Weight 219.0273
Percent Purity >95%
Chemical Name or Material FR 900098 Monosodium Salt
Grade HPLC