missing translation for 'onlineSavingsMsg'
Learn More

FOXR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. p-7209772 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16851495 20 μL 20µL
16841495 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16851495 Supplier Proteintech Supplier No. 219421AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The forkhead-box (FOX) genes comprise a superfamily of at least 43 members that express proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 amino acids. FOX transcription factors play roles in determining cell fate during early development. FOXR1 (forkhead box protein R1), also known as FOXN5 (forkhead box protein N5) or DLNB13, is a 292 amino acid nuclear protein that belongs to the FOX family and contains one forkhead DNA-binding domain. Through chromosomal aberrations such as retroviral integration, gene amplification or translocation, FOXR1 may be involved the development of certain invasive carcinomas.

Specifications

Antigen FOXR1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FOXR1
Gene Accession No. Q3UTB7, Q6PIV2
Gene Alias DLNB13, Forkhead box protein N5, Forkhead box protein R1, forkhead box R1, FOXN5, FOXR1
Gene Symbols FOXR1
Host Species Rabbit
Immunogen FOXR1 Fusion Protein Ag16625
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283150, 382074
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.