missing translation for 'onlineSavingsMsg'
Learn More

Fosmidomycin sodium salt, MedChemExpress

Product Code. 16417437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417437 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417437 Supplier Medchem Express Supplier No. HY112853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fosmidomycin sodium salt is a phosphonic acid antibiotic and a antimalarial drug, which is active against both Gram-negative and Gram-positive bacteria.

Fosmidomycin sodium salt is a phosphonic acid antibiotic and a antimalarial drug, which is active against both Gram-negative and Gram-positive bacteria.

Specifications

Chemical Name or Material Fosmidomycin sodium salt
Physical Form Solid
CAS 66508-37-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C4H9NNaO5P
For Use With (Application) Neuroscience-Neuromodulation
Synonym FR-31564
Solubility Information H2O : 25 mg/mL (121.90 mM; ultrasonic and warming and heat to 60°C)
SMILES OP(CCCN(C=O)O)(O[Na])=O
Molecular Weight (g/mol) 205.08
Formula Weight 205.08
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.