Learn More
Fosamprenavir Calcium Salt, TRC
Product Code. 30390741
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390741 10 mg 10mg
30402071 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30390741 Supplier Toronto Research Chemicals Supplier No. TRCF72725010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 226700-81-8
Molecular Formula C25 H34 N3 O9 P S . Ca
Molecular Weight (g/mol) 623.67
Synonym Carbamic acid, N-[(1S,2R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-1-(phenylmethyl)-2-(phosphonooxy)propyl]-, (3S)-tetrahydro-3-furanyl ester, calcium salt (1:1), Carbamic acid, [(1S,2R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-1-(phenylmethyl)-2-(phosphonooxy)propyl]-, C-[(3S)-tetrahydro-3-furanyl] ester, calcium salt (1:1) (9CI), Fosamprenavir calcium, GW 433908G, Lexiva, GW 433908G, fosamprenavir calcium pentahydrate
IUPAC Name calcium;[(2R,3S)-1-[(4-aminophenyl)sulfonyl-(2-methylpropyl)amino]-3-[[(3S)-oxolan-3-yl]oxycarbonylamino]-4-phenylbutan-2-yl] phosphate
SMILES [Ca+2].CC(C)CN(C[C@@H](OP(=O)([O-])[O-])[C@H](Cc1ccccc1)NC(=O)O[C@H]2CCOC2)S(=O)(=O)c3ccc(N)cc3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C25H36N3O9PS.Ca/c1-18(2)15-28(39(33,34)22-10-8-20(26)9-11-22)16-24(37-38(30,31)32)23(14-19-6-4-3-5-7-19)27-25(29)36-21-12-13-35-17-21;/h3-11,18,21,23-24H,12-17,26H2,1-2H3,(H,27,29)(H2,30,31,32);/q;+2/p-2/t21-,23-,24+;/m0./s1
Formula Weight 623.1379
Chemical Name or Material Fosamprenavir calcium salt