Learn More
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser
Click to view available options
Capacity (Metric):
30 mL
50 mL
Unit Size:
Each
Description
- No metal spring
- No contamination of the dosing liquid
- According to EN ISO 8655-5
- Made of PTFE
Specifications
Specifications
| Quantity | 1 |
| Type | Bottle Top Dispenser |
| Capacity (Metric) | 50 mL |