Learn More
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser is for dispensing hydroflouric acid and all liquids that can be handled with the OPTIFIX™ dispensers.
Brand:  Poulten & Graf GmbH 101 10644
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10272941
  • 1410.00 EUR / Each
Estimated Shipment: 17-06-2024
Description
  • No metal spring
  • No contamination of the dosing liquid
  • According to EN ISO 8655-5
  • Made of PTFE
Specifications
Specifications
30 mL
1
Bottle Top Dispenser