Learn More
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser is for dispensing hydroflouric acid and all liquids that can be handled with the OPTIFIX™ dispensers.
1410.00 EUR - 2115.00 EUR
Specifications
Quantity 1
Type Bottle Top Dispenser
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11758286
View Documents Promotion Details
Poulten & Graf GmbH
101 10648
50 mL
2115.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
10272941
View accessory products View Documents Promotion Details
Poulten & Graf GmbH
101 10644
30 mL
1410.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
Description
  • No metal spring
  • No contamination of the dosing liquid
  • According to EN ISO 8655-5
  • Made of PTFE
Specifications
Specifications
1
Bottle Top Dispenser