Learn More
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser is for dispensing hydroflouric acid and all liquids that can be handled with the OPTIFIX™ dispensers.
1410.00 EUR - 2115.00 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11758286
|
Poulten & Graf GmbH
101 10648 |
50 mL |
2115.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10272941
|
Poulten & Graf GmbH
101 10644 |
30 mL |
1410.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- No metal spring
- No contamination of the dosing liquid
- According to EN ISO 8655-5
- Made of PTFE
Specifications
1 | |
Bottle Top Dispenser |