missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 85% w/w, Extra Pure, Scharlab
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
HDPE Carboy
Unit Size:
1 L
5 L
Description
Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.
Specifications
Specifications
| Melting Point | -9°C |
| Boiling Point | 107°C |
| Color | Colorless |
| Linear Formula | HCOOH |
| UN Number | 3412 |
| Solubility Information | Miscible in water |
| Formula Weight | 46.03g/mol |
| Percent Purity | 85% (min.) |
| Grade | Applications: analytical chemistry, synthesis of organic products, in the rubber industry, acidifying agent, cosmetics |
| Density | 1.2g/cm3 |
| Show More |
Safety and Handling
Product Identifier
- Formic acid, solution 85% w/w, extra pure
- Danger
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Formic Acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction