missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 4% Buffered, Solveco

Product Code. 30281148
Change view
Click to view available options
Packaging:
10 L Bag in a Box
10 L Container
Unit Size:
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
30281148 10 L Container 10 L
30281149 10 L Bag in a Box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30281148 Supplier Solveco Chemicals Supplier No. 1724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formaldehyde 4% (Formalin 10%) saline-buffered for use in tissue sample fixation.

Formaldehyde 4% is a high-quality fixative solution specifically formulated for use in histopathology. This solution is ideal for preserving tissue specimens, ensuring their integrity and stability for accurate microscopic examination.

  • Effective Tissue Fixation: The 4% concentration of formaldehyde is optimized for halting enzymatic and bacterial degradation after tissue removal, preserving cellular and extracellular structures as closely as possible to their natural state.
  • Buffered Formulation: Buffered with salt, this solution maintains a stable pH level, which is crucial for preventing the formaldehyde from becoming acidic over time. This buffering ensures consistent fixation results and prevents potential degradation of the tissue sample.
  • Maintains Tissue Architecture: Cross-links proteins in the tissue, stabilizing cellular morphology and composition for reliable histological analysis.
  • Ease of Sectioning: Fixed tissues become firm and easy to cut into thin sections, facilitating smooth preparation for microscopic examination.
  • Long-term Storage: Provides excellent preservation, allowing tissue samples to be stored for extended periods without significant degradation.
  • Enhanced Staining: Ensures effective staining, making it easier to visualize and analyze different tissue components under the microscope.

Formaldehyde 4% is essential for accurate tissue fixation and reliable histopathological results, making it a staple in laboratory settings.

The product is CE-marked as a general IVD product in accordance with the In Vitro Diagnostic Directive (Regulation (EU) 2017/746 on in vitro diagnostic medical devices).

Specifications

Packaging 10 L Container
Quantity 10 L
Molecular Weight (g/mol) 30.03 g/mol
Percent Purity 4%
Grade Reagent
Physical Form Viscous Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.