missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, 37% w/w, Stabilized with Methanol, Reagent Grade, Scharlab
Description
- Synonyms: Formalin solution, Formol, Methanal solution, Methyl aldehyde solution
- Molecular Formula: CH2O
- Molar Mass: 30.03g/mol
- CAS: 50-00-0
- EIN ECS No.: 200-001-8
- Ignition Temperature: ∼300°C (pure substance)
- Vapor Pressure: 1.3hPa (formaldehyde)
Specifications
Specifications
| Melting Point | <-15°C |
| Density | 1.09g/cm3 |
| Boiling Point | 93°C to 96°C |
| Flash Point | 62°C |
| Ignition Residue | 0.002% max |
| Quantity | 2.5 L |
| Stabilizer | Approx. 10% methanol |
| Formula Weight | 30.03g/mol |
| Grade | Reagent |
| Concentration or Composition (by Analyte or Components) | 37% (acidimetric assay) |
| Show More |
Safety and Handling
Product Identifier
- Formaldehyde, solution 37% w/w, reagent grade, stabilized with approx. 10 % methanol
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H311 + H331-Toxic in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Formaldehyde, Methanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?