missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37% w/w, Stabilized with Methanol, Reagent Grade, Scharlab

Product Code. 11304134
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304134 1 L 1 L
11314134 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304134 Supplier Scharlab Supplier No. FO00111000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a disinfectant and in the synthesis of organic products, embalming liquids, analytical chemistry, photography and the pharmaceutical industry.

  • Synonyms: Formalin solution, Formol, Methanal solution, Methyl aldehyde solution
  • Molecular Formula: CH2O
  • Molar Mass: 30.03g/mol
  • CAS: 50-00-0
  • EIN ECS No.: 200-001-8
  • Ignition Temperature: ∼300°C (pure substance)
  • Vapor Pressure: 1.3hPa (formaldehyde)

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point <-15°C
Density 1.09g/cm3
Boiling Point 93°C to 96°C
Flash Point 62°C
Ignition Residue 0.002% max
Quantity 1 L
Stabilizer Approx. 10% methanol
Formula Weight 30.03g/mol
Grade Reagent
Concentration or Composition (by Analyte or Components) 37% (acidimetric assay)
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, solution 37% w/w, reagent grade, stabilized with approx. 10% methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.