Learn More
Thermo Scientific™ Forma™ Inner Door Kits
Product Code. 11386341 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Forma 7000 and 900 Series, 17.3 cu. ft. Uprights
Forma 7000 and 900 Series, 23.0 cu. ft. Uprights
Forma 7000 and 900 Series, 28.0 cu. ft. Uprights
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11386341 Forma 7000 and 900 Series, 23.0 cu. ft. Uprights Each
11376341 Forma 7000 and 900 Series, 17.3 cu. ft. Uprights Each
11386331 Forma 7000 and 900 Series, 28.0 cu. ft. Uprights Each
3 options
Product Code. 11386341
Brand: Thermo Scientific™ 1950548
Only null left
Add to basket
Add to basket
Reduce cold air loss and improve temperature recovery after door closings when you choose factory-installed Thermo Scientific™ Forma™ Inner Door Kits for your ultra-low upright freezer.
Reduce cold air loss and improve temperature recovery after door closings with Thermo Scientific™ Forma™ Inner Door Kits, a factory-installed door alternative option for your ultra-low upright freezer.
Four interior doors are already included on our -86°C uprights. The five inner door option for 8600 and 900 Series includes five powder-coated inner doors and one additional shelf.
Ordering Information:
Factory installed. Must be ordered with the refrigerator or freezer.
TRUSTED_SUSTAINABILITY
Specifications
Type Ultra-Low Freezer Inner-Door
Capacity (English) 23 cu. ft.
Capacity (Metric) 651.2 L
Temperature Range -86°C
No. of Doors 5
Exterior Finish Powder-coated Steel
Shelves 1
Style Upright
For Use With (Equipment) Forma 7000 and 900 Series, 23.0 cu. ft. Uprights