Learn More
Thermo Scientific™ Forma™ 8600 Series -40°C Ultra-Low Temperature Chest Freezers
Store samples confidently in the Forma 8600 Series -40°C chest freezer. Available in four sizes to accomodate a variety of needs.
POA
Specifications
Type | Ultra Low Freezer, Chest |
---|---|
Temperature Range | -10°C to -40°C |
Cabinet Material | Cold Rolled Steel |
Insulation | Polyurethane Foam |
Interior | Stainless Steel |
Description
Store samples confidently in Thermo Scientific™ Forma™ 8600 Series -40°C ultra-low temperature chest freezers. Available in four sizes, and with a full range of racking solutions to accomodate a variety of needs, these freezers deliver all of the same sample protection features of the Forma™ 900 and 7000 upright series.
Key Features:
- Centralized information center, including microprocessor control and monitoring system ensures that all controls and displays are easy to reach and read—knee level on the 3 cu. ft. models
- Improves temperature control and increases reserve BTUH capacity, resulting in a more stable temperature sample
- Easy to remove, washable filter provides protection from dust on the condenser, causing reduced refrigeration performance and increased risk to samples
- Simplified installation with our new easy-roll 2 inch locking casters
- Easy to use ergonomic door handle allows convenient disposal, and retieval of, specimens
Construction:
- 5 in.; (127mm) foamed in place, polyurethane insulation reduces power consumption and maintains temperature setpoint
- Triple-point door gasket provides longer holding time in the event of a door opening
- Rugged, heavy gauge, cold-rolled steel cabinets resist chipping and rust
- 1 in. (25mm) access port allows for the use of inexpedient probes or instrumentation
- Stainless steel interior
- Polystyrene interior sub-lids reduce cold air loss and improve temperature recovery after door openings
Warranty varies by country, please contact us for details
Specifications
Ultra Low Freezer, Chest | |
Cold Rolled Steel | |
Stainless Steel | |
Chest | |
Fin and Tube, Forced-Air Cooled | |
Forma | |
Manual | |
Alarm, Recorder | |
Optional 7-day chart recorder | |
Standard Operation | |
CO2 or LN2 Optional | |
970 W | |
Hermetic Compressor |
-10°C to -40°C | |
Polyurethane Foam | |
CE, cULus, FDA Class I, UL | |
Push Button LED | |
Push Button LED | |
Warranty varies by country, please contact us for details | |
Forma | |
Key Lock | |
RS-485, 4-20mA, dry contacts - standard | |
Single ID | |
Smart-Vue Compatible | |
Buck/Boost | |
R404A |