Description
- Corrugated handles
- Serrated tips
- Disposable
Specifications
Specifications
| Tip Style | Straight |
| Design | Serrated Tips |
| Length (English) | 4.4 in. |
| Material | Plastic |
| Length (Metric) | 11.1 cm |
| Product Type | Forceps |
| Quantity (No. per Case) | 6 Pk. |
| Includes | Corrugated handles |
| Type | Disposable Forceps |
| Style | High-precision |