Learn More
Memmert™ Flush-Fit Sets without Air Slots for UF750plus
Product Code. 17509585 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF750plus Oven
Förpackningsstorlek
Each
Denne vare kan ikke returneres. Se returpolitik
Produktkode 17509585
missing translation for 'mfr': Memmert™ UF750plusB29743
Kun null tilbage
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Stainless steel frame covering gap between oven and wall opening.
  • Stainless steel
  • Without air slots
  • Techincal clarification required
Specifications
For Use With (Equipment) UF750plus Oven
Type Flush-fit Unit
Material Stainless Steel