missing translation for 'onlineSavingsMsg'
Learn More

Fluoroglycofen-ethyl, TRC

Product Code. 30391773
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391773 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391773 Supplier Toronto Research Chemicals Supplier No. TRCF621120500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluoroglycofen-ethyl
CAS 77501-90-7
Molecular Formula C18 H13 Cl F3 N O7
Quantity 500 mg
Synonym Benzoic acid, 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitro-, 2-ethoxy-2-oxoethyl ester, Compete, Fluoroglycofen-ethyl, RH 0265, Simtar, 2-Ethoxy-2-oxoethyl 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoate
InChI Formula InChI=1S/C18H13ClF3NO7/c1-2-28-16(24)9-29-17(25)12-8-11(4-5-14(12)23(26)27)30-15-6-3-10(7-13(15)19)18(20,21)22/h3-8H,2,9H2,1H3
SMILES CCOC(=O)COC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]
IUPAC Name (2-ethoxy-2-oxoethyl) 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoate
Molecular Weight (g/mol) 447.75
Formula Weight 447.0333
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.