Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Fluoride Buffer Solution |
| Name Note | According to DEV D4 |
| CAS | 7732-18-5, 145819-99-4, 6132-04-3 |
| Quantity | 1 L |
| UN Number | NONH for all modes of transport |