Learn More
Fluorescein disodium salt, Thermo Scientific™
Fluorescein disodium salt (uranin, fluorescite, acid yellow 73, CAS # 518-45-6) is the salt of the xanthene dye commonly used for fluorescent tracing.
37.65 EUR - 83.65 EUR
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464139
SDS View Documents
Thermo Scientific Alfa Aesar
J61549.22
ONLINE EXCLUSIVE PRICE
42.20 EUR
37.65 EUR / 100g
Save 4.55 EUR
10% Off
View alternative products
Save Up to 
»
15474139
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J61549.36
500g
ONLINE EXCLUSIVE PRICE
93.30 EUR
83.65 EUR / 500g
Save 9.65 EUR
10% Off
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Fluorescein disodium salt is a yellow-green phthalate indicator dye that is pH-sensitive
• It has a maximum absorbance of 494nm and maximum emission of 521nm
Application
• Fluorescein disodium salt can be used as a fluorescent protein labeler
• Fluorescein is also useful for the staining of cells and organelles