Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Product Code. 17890567 Shop All Thermo Scientific Products
Click to view available options
Tubing Size:
TPE 374, 1 m (3.25 ft)
TPE 374, 2.5 m (8.2 ft)
TPE 374, 5 m (16.4 ft)
Connector Types:
Luer
MPC
MPX
Dimensions:
1.2 in. ID x 3/4 in. OD
1/4 in. x 7/16 in. OD
1/8 in. ID x 1/4 in. OD
3/8 in. ID x 5/8 in. OD
Unit Size:
Pack of 10
Product Code. 17890567
Brand: Thermo Scientific™ SH31119.01
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
  • Irradiation dose of 25-40 kGy provides minimum SAL of 10-6 for product contact surfaces
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 1/8 in. ID x 1/4 in. OD
Tubing Style 3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 1 m (3.25 ft); Luer
Connector Types Luer
Product Type Extension Set
Tubing Size TPE 374, 1 m (3.25 ft)