Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Product Code. 17830657 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
Luer to MPC
MPC/MPX
MPX to MPC
MPX/MPC
Tubing Size:
C-Flex 374, 2 m (6.6 ft)
C-Flex 374, 2 m (78.7 in)
TPE 374, 1 m (3.28 ft)
TPE 374, 2 m (6.6 ft)
TPE 374, 2 m (78.7 in)
Unit Size:
Pack of 10
Product Code. Connector Types Tubing Size unitSize
17830657 MPC/MPX TPE 374, 2 m (78.7 in) Pack of 10
17850577 Luer to MPC TPE 374, 1 m (3.28 ft) Pack of 10
17870647 Luer to MPC TPE 374, 2 m (6.6 ft) Pack of 10
17830577 MPX to MPC TPE 374, 2 m (78.7 in) Pack of 10
17870577 MPX to MPC TPE 374, 2 m (6.6 ft) Pack of 10
17860577 MPX to MPC C-Flex 374, 2 m (78.7 in) Pack of 10
17890647 MPX to MPC C-Flex 374, 2 m (6.6 ft) Pack of 10
17800587 MPX/MPC TPE 374, 2 m (78.7 in) Pack of 10
17800657 MPX/MPC TPE 374, 2 m (6.6 ft) Pack of 10
17840657 MPC/MPX TPE 374, 2 m (6.6 ft) Pack of 10
10 options
Product Code. 17830657
Brand: Thermo Scientific™ SH31129.01
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifications
Tubing Style 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (78.7 in); MPX to MPC
Connector Types MPC/MPX
Product Type Adapter Set
Tubing Size TPE 374, 2 m (78.7 in)