missing translation for 'onlineSavingsMsg'
Learn More

Flugestone 17-Acetate, TRC

Product Code. 18012850
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012850 25 mg 25mg
18083140 10 mg 10mg
18071250 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18012850 Supplier Toronto Research Chemicals Supplier No. TRCF48965025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2529-45-5
Molecular Formula C23 H31 F O5
Molecular Weight (g/mol) 406.49
Synonym Pregn-4-ene-3,20-dione, 9-fluoro-11β,17-dihydroxy-, 17-acetate (7CI,8CI), Progesterone, 9-fluoro-11β,17-dihydroxy-, 17-acetate (6CI), (11β)-17-(Acetyloxy)-9-fluoro-11-hydroxypregn-4-ene-3,20-dione, 17α-Acetoxy-9α-fluoro-11β-hydroxy-pregn-4-ene-3,20-dione, 17α-Acetoxy-9α-fluoro-11β-hydroxyprogesterone, 9α-Fluoro-11β-hydroxy-17α-acetoxypregn-4-ene-3,20-dione, Chronogest, Cronolone, FGA, Flugestone acetate, Fluorogestone acetate, Flurogestone acetate, Gyncro-Mate, NSC 65411, SC 9880, Synchronate, Syncro-Mate
IUPAC Name [(8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H31FO5/c1-13(25)22(29-14(2)26)10-8-17-18-6-5-15-11-16(27)7-9-20(15,3)23(18,24)19(28)12-21(17,22)4/h11,17-19,28H,5-10,12H2,1-4H3/t17-,18-,19-,20-,21-,22-,23-/m0/s1
Formula Weight 406.2156
Chemical Name or Material Flugestone 17-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.