Learn More
Masterflex™ Flow Tubes
Click to view available options
Airflow Rate:
2.5S CFH
25 mL/min.
Unit Size:
Each
Specifications
Specifications
| Type | Flowtube |
| Accuracy | ±2% (Full Scale) |
| Airflow Rate | 2.5S CFH |
| Pressure Rating | 200 psig |
| Connections | 1/8 in. NPT (F) |
| For Use With (Application) | Direct-Reading Flowtube for Air |
| Length (Metric) | 150 mm |
| Repeatability | ±0.25% |
| Temperature (Metric) | 121°C |