Learn More
Axygen™ FLIPR™ Liberty Robotic Tips
Accurate sample delivery
Brand:  Axygen™ LT-384-BK-R
Additional Details : Weight : 10.14000kg
 View more versions of this product
Product Code. 10804571
  • 4215.00 EUR / Pack of 19200
Estimated Shipment: 01-11-2024
Description
  • Available in clear and black
  • Volume: 30μL
  • Available as racked or racked and pre-sterilized
  • For use with Molecular Devices Liberty head
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.
Specifications
Specifications
30 μL
Black
DNase, RNase, and Pyrogen free
Automation, Round
50 Racks/Cs.
FLIPR
Non-sterile
Autoclavable
Rack
SBS Rack
Molecular Devices FLIPR 384-well Head