Learn More
Cole-Parmer™ Single Flint Renewal
Shop All Cole Parmer Products
Click to view available options
For Use With (Equipment):
Safety Spark Lighter
Unit Size:
Pack of 5
Description
- No fuel to change or store.
- It is seated in the aluminum flint holder.
- Larger size for longer life.
- Compatible for use with the Safety Spark Lighter, model 36136-50.
- Striker sold separately.
Specifications
Specifications
| Product Type | Replacement Flint |
| For Use With (Equipment) | Safety Spark Lighter |
| No. per Pack | 5 Pk. |