Learn More
Testo™ Flexible Oven Probe
Product Code. 10074411
Click to view available options
Temperature Range (Metric):
-50°C to +250°C
Unit Size:
Each
Product Code. 10074411
Brand: Testo™ 0603 0646
Only null left
Specifications
Includes Flexible Oven Probe, 1.5 dia. x 2000mm Probe Shaft, –50 to 250°C Measuring Range, Thermocouple Type T, Heat Resistant Probe Cable
Certifications/Compliance EN 13485 and HACCP-compliant
Temperature Range (Metric) -50°C to +250°C