Learn More
JULABO™ Flexible Metal Tubing
Product Code. 15234323
Change view
Click to view available options
Connection Type:
2 fittings M24 x 1.5 female
2 fittings M30 x 1.5 female
2 fittings M38 x 1.5 female
Size:
1 m
1.5 m
2 m
3 m
5 m
Unit Size:
Each
Pack of 1.5
Pack of 2
Pack of 3
Pack of 5
Product Code. Connection Type Size unitSize
15234323 2 fittings M30 x 1.5 female 3 m Pack of 3
15274313 2 fittings M24 x 1.5 female 1.5 m Pack of 1.5
15264313 2 fittings M24 x 1.5 female 1 m Each
15284313 2 fittings M24 x 1.5 female 2 m Pack of 2
15204323 2 fittings M30 x 1.5 female 1 m Each
15214323 2 fittings M30 x 1.5 female 1.5 m Pack of 1.5
15224323 2 fittings M30 x 1.5 female 2 m Pack of 2
15244323 2 fittings M30 x 1.5 female 5 m Pack of 5
15254323 2 fittings M38 x 1.5 female 1.5 m Pack of 1.5
15264323 2 fittings M38 x 1.5 female 2 m Pack of 2
15274323 2 fittings M38 x 1.5 female 3 m Pack of 3
15284323 2 fittings M38 x 1.5 female 5 m Pack of 5
12 options
Product Code. 15234323
Brand: JULABO™ 8930274
Only null left
Add to basket
Add to basket
For use with PRESTO™ Temperature control system/process system. Triple insulated for use up to 350°C.
  • Flexible Metal tubing
  • Triple insulated
  • -100 ... +350 °C.
Available lengths and fittings:
  • 1 m:
    • M24 x 1.5 female
    • M30 x 1.5 female
  • 1.5 m:
    • M24 x 1.5 female
    • M30 x 1.5 female
    • M38 x 1.5 female
  • 2 m:
    • M24 x 1.5 female
    • M30 x 1.5 female
    • M38 x 1.5 female
  • 3 m:
    • M30 x 1.5 female
    • M38 x 1.5 female
  • 5 m:
    • M30 x 1.5 female
    • M38 x 1.5 female
Specifications
Description Metal tubing
For Use With (Equipment) JULABO™ PRESTO™ A45, PRESTO™ A45t, PRESTO™ W50, PRESTO™ W50t, PRESTO™ W55, PRESTO™ A85, PRESTO™ A85t
Temperature Range -100°C to +350°C
Material Metal
Size 3 m
Connection Type 2 fittings M30 x 1.5 female