Learn More
Memmert™ Flat Stainless Steel Cover with Openings and Concentric Ring Sets
Shop All Memmert Products
Click to view available options
Quantity:
3 Pk.
6 Pk.
8 Pk.
For Use With (Equipment):
WTB15
WTB35
WTB50
WTB6
Unit Size:
Each
Description
- Accessory
- Wwith openings and concentric ring sets
Specifications
Specifications
| For Use With (Equipment) | WTB15 |
| Type | Flat stainless steel cover |
| Material | Stainless Steel |
| Quantity | 6 Pk. |
| Diameter (Metric) Min. Opening | 107 mm |