Learn More
Labnet™ Flat Platform for Orbit™ 1900 Digital Heavy Duty Shaker
Product Code. 11426727
Click to view available options
For Use With (Equipment):
Orbit™ 1900 Digital Heavy Duty Shaker
Unit Size:
Each
Product Code. 11426727
Brand: Labnet™ S204012
Orbit™ 1900 Digital Heavy Duty Shaker Flat Platform is useful for shaking trays, plates, boxes and other containers.
  • The Orbit™ 1900 is a large shaker designed for continuous duty and heavy loads. Its solid construction and counter balanced drive system provide vibration free operation.
    • Flat or flask clamp platform options
    • Designed for heavy loads and continuous duty
    • Safe for use in temperature controlled environments
    • Ideal for liquid cultures and general mixing
  • Operation of the shaker can be timed or continuous, while the speed is variable. Both time and speed are set digitally. Setting parameters is easy with the analog knob. Values are shown on the large, three digit display.
  • The flat platform is useful for shaking trays, plates, boxes and other containers. A nonslip mat on the platform surface keeps items in place.
Specifications
Product Type Flat Platform
For Use With (Application) Shaking Trays, Plates, Boxes and Other Containers.
For Use With (Equipment) Orbit™ 1900 Digital Heavy Duty Shaker
Includes Non-Slip Mat