Description
- Manufactured from borosilicate glass 3.3 ground, component for further processing into reaction vessels
- Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications,glass articles for further processing
- Available with or without grooves, they deliver reliable sealing, high mechanical strength, and trusted DURAN™ quality for demanding laboratory use.
Specifications
Specifications
| Product Type | Flat Flange |
| For Use With (Equipment) | Pressure‐resistant Reaction Vessels |
| Includes | Without Groove |
| Material | Borosilicate Glass |
| Quantity | 1 Ea. |