Learn More
Agencinox™ Flat Bottom Stainless Steel Dishes
Click to view available options
Capacity (Metric):
60 mL
100 mL
250 mL
Unit Size:
Each
Description
- Flat bottom stainless steel lotion bowls
- Autoclavable
- Available in a variety of sizes
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 60 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Lotion Bowl |
| Height (Metric) | 30 mm |
| Diameter (Metric) | 60 mm |