Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Narrow Neck
Product Code. p-7222606
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
4 L
5 L
10 L
Height (Metric):
90 mm
105 mm
138 mm
163 mm
190 mm
250 mm
259 mm
275 mm
290 mm
360 mm
Unit Size:
Each
Pack of 10
Product Code. 10080401
Brand: Lenz Laborglasintrumente 07045070
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Capacity (Metric) 1 L
Material Duran Glass
Height (Metric) 190 mm