Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07045037
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10409562
  • 146.28 EUR / Pack of 10
Estimated Shipment: 25-06-2024
Description
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
100 mL
Duran Glass
105 mm