Learn More
Fisherbrand™ Flask Clamp for Shaking Incubators
Shop All Fisherbrand Products
Click to view available options
Holds:
1000 mL Flasks
125 mL Flasks
125, 250, 500 mL Flasks
25 mL Flasks
250 mL Flasks
50 mL Flasks
500 ml Flasks
Unit Size:
Each
Specifications
Specifications
| Holds | 1000 mL Flasks |
| Product Type | Flask Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 16313226 |
| Packaging Quantity | 1 Ea. |