Learn More
Flash Point Standard, PMCC ASTM D 93 Proc A (nominal value 228.0 °C), Paragon Scientific™
Product Code. 16050040
Change view
Click to view available options
Quantity:
3 x 80 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16050040 3 x 80 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16050040 Supplier Paragon Scientific Supplier No. CRMUPMCCHIGH
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D93 Procedure A. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard
  • Certified Reference Material for annual verification checks in accordance with ASTM D93, Procedure A.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom.
Specifications
Quantity 3 x 80 mL
Nominal values quoted. Certified Values will vary from batch to batch.