Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Product Code. 17864961 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
5 μL
10 μL
50 μL
100 μL
250 μL
Needle Gauge:
23 G
23 to 26s G
26 G
31 G
Quantity:
6 Pk.
Ea.
Needle Point Style:
Cone
Side Hole
missing translation for 'unitSize'
Each
Pack of 6
Product Code. 17864961
missing translation for 'mfr': Thermo Scientific™ 365G1503
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 G
Product Type Gas Tight GC Autosampler Syringe, Removable Needle
Quantity Ea.
Volume (Metric) 50 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) AI/AS 1610 Autosampler
Connection Type Removable
Replacement Needle 36550040 (Side Hole), 36566485 (Cone)
PTFE Tipped Plunger Yes