Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
730.00 EUR - 1045.00 EUR
Specifications
Type | Fixed Angle Rotor |
---|---|
For Use With (Equipment) | 5430/5430R Centrifuges |
Angle | Fixed |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
13477916
|
Eppendorf™
5427739009 |
6 x 15/50 mL |
1045.00 EUR
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13276928
|
Eppendorf™
5427752005 |
24 Spin Columns |
935.00 EUR
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13286928
|
Eppendorf™
5427753001 |
30 x 1.5/2 mL |
730.00 EUR
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13206938
|
Eppendorf™
5427754008 |
48 x 1.5 mL |
850.00 EUR
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Fixed Angle Rotor | |
5430/5430R Centrifuges | |
Fixed |