Learn More
Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 506/780 Sample Pack
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
- Fixable Viability Dyes may be used to label cells from all species
- Supplied as pre-diluted solutions prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture
- Store at −70°C with dessicant
- It may be freeze-thawed up to 20 times
- Allow vial to equilibrate to room temperature before opening
- Can be excited by the violet (405nm) laser line and has a peak emission of 506nm
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- Can be excited by the red (633nm) laser line and has a peak emission of 780nm
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- Staining may be done before or after surface staining
Specifications
Specifications
| Color | Green, Infrared |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Dye |
| Emission | 405/506, 633/780 |
| Product Line | eBioscience |
| Quantity | 1 kit |