missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FITC Monoclonal Antibody (#9)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA514696

Product Code. 11539281

  • 544.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product MA514696 is a smaller package size of MIF2902 (formerly sold as a Seradyn product).

FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FITC
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Mouse
Protein A
RUO
Chemical
Antibody
IgG2a λ
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
#9
Unconjugated
Fluorescein
FITC-BSA.
100 μg
Primary
Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.