Learn More
Fisherbrand™ Polymethylpentene Graduated Cylinders
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
20 mL
missing translation for 'unitSize'
Pack of 12
Pack of 18
Pack of 4
Pack of 6
Pack of 8
Description
Calibrated To Contain/To Deliver at 20°C
- Transparent cylinders with large molded-in graduations
- Easy to read
- Large rounded bases ribbed for reinforcement and stability
- Generous pour spout; 10mL size has a conical top for easier filling and pouring
- Non-wetting
- Chemical, heat and impact resistant
- Window-clear
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | PMP |
| Calibration | To Contain, To Deliver |
| Graduations | 20 mL |
| Autoclavable | Autoclavable |
| Subdivision | 20mL |
| Base Type | Circular |
| Type | Graduated Cylinder |