Learn More
Stopper,red rubber,2 hole diameter 86mm
Brand:  Fisherbrand RB086/2H
Stopper,red rubber,2 hole diameter 86mm
Product Code. 10020351
  • ONLINE EXCLUSIVE PRICE
    32.55 EUR
    29.04 EUR / Each
    Save 3.51 EUR
    10% Off
Only null left
Estimated Shipment: 18-03-2026