Learn More
Stopper red rubber d=86mm one hole
Brand:  Fisherbrand RB086/1H
Stopper red rubber d=86mm one hole
Product Code. 10161801
  • ONLINE EXCLUSIVE PRICE
    32.55 EUR
    29.04 EUR / Each
    Save 3.51 EUR
    10% Off
Only null left
Estimated Shipment: 26-02-2026