Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 1.5mm Thick
Product Code. 11837813 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
18
30
36
48
5
60
Volume (Metric) Well:
30 μL
40 μL
50 μL
60 μL
80 μL
160 μL
320 μL
2200 μL
Unit Size:
Each
Product Code. 11837813
Only null left
Add to basket
Add to basket
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications
Color Red
For Use With (Equipment) Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
Type Comb
No. of Wells 10
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Well 160 μL