Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 0.75mm Thick
Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
18
24
30
36
48
5
Volume (Metric) Well:
15 μL
20 μL
25 μL
30 μL
40 μL
80 μL
160 μL
1100 μL
Unit Size:
Each
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units |
| Type | Comb |
| No. of Wells | 30 |
| Quantity | 1 Each |
| Thickness (Metric) | 0.75 mm |
| Volume (Metric) Well | 25 μL |