Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 0.75mm Thick
Product Code. 11807813 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
18
24
30
36
48
5
Volume (Metric) Well:
15 μL
20 μL
25 μL
30 μL
40 μL
80 μL
160 μL
1100 μL
Unit Size:
Each
Product Code. 11807813
Only null left
Add to basket
Add to basket
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications
Color Black
For Use With (Equipment) Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
Type Comb
No. of Wells 1 Prep, 1 Marker
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Well 1100 μL