Learn More
Fisherbrand™ 96-Well Nonskirted PCR Plates
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Unit Size:
Pack of 25
Description
Compatible with most 0.2mL thermal cyclers and ABI PRISM™ 310, 3100, and 3700 capillary sequencers
- Virgin polypropylene
- Clear well bottom for sample visibility
- Highly rigid plate deck
- Raised rim design
- Sealable using adhesive films and foils, heat seals, and cap strips
- 300μL maximum volume when used with adhesive and heat seals
- Cut-away corner and alphanumeric grid
- Cleanroom produced
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Green |
| Product Type | PCR Microplate |
| Skirt Style | Non-skirt |
| Material | Polypropylene |
| Volume (Metric) Well | 300 μL |
| No. of Wells | 96 |
| For Use With (Application) | PCR, RT-PCR applications |
| Rimmed | Yes |
| Show More |