Learn More
Fisherbrand™ Traceable™ DAS-4 Data Logger System
Shop All Fisherbrand Products
Click to view available options
Data Memory:
8000 readings
Unit Size:
Each
Description
Captures and stores up to 8000 readings
- Readings may be taken at intervals from 1 second to 99 hours
- Stored readings may be downloaded to any IBM-compatible computer
- Data files can be read by spreadsheets, databases, and statistical programs
Order Info
Supplied with 36 in. (91.4cm) serial cable with D9F plug, 3.50 in. diskette, and four AA alkaline batteries. Unit measures 1L x 3W x 5 in. H (2.5 x 7.6 x 12.7cm); weighs 7 oz. (198.4g)
Specifications
Specifications
| Type | Datalogger System |
| Data Memory | 8000 readings |
| Calibration | Not Calibrated |
| For Use With (Application) | Captures and stores up to 8000 readings |