Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
Click to view available options
Volume (Metric):
500 μL
1250 μL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 25
Pack of 50
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
- Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Specifications
| Volume (Metric) | 1250 μL |
| Volume (Metric) Dispensing | 25, 50, 75, 100, 125 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Certifications/Compliance | ISO 9001, ISO 14001 |
| Format | Bulk |
| For Use With (Equipment) | Thermo Scientific FinnPipette Stepper |
| Product Line | FinnTip |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion