Learn More
Brady™ Fine Pen
Click to view available options
Color:
Black
Unit Size:
Each
Description
Specially formulated to resist oil, abrasion and fading.
Specifications
Specifications
| Tip Style | Fine |
| Alternate Part Number | BFS-05 |
| Product Type | Permanent Marker |
| Color | Black |
| Description | Pen - Fine |
| For Use With (Application) | Used For Marking Laboratory Glass Wares |
| Quantity | 1 Pk. |